Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691245.3 | XP_005691302.1 | 1 - 2 | 2913 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |