Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061757.1 | XP_017917246.1 | 2 - 3 | 273 | 102 | |
GenBank mRNA | JO361973.2 | - | 2 - 3 | 440 | 102 | |
JT760161.1 | - | 2 - 3 | 170 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |