Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064030.1 | XP_017919519.1 | 8 - 9 | 1215 | 58 | |
GenBank mRNA | JO562412.2 | - | 4 - 5 | 517 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |