Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696286.3 | XP_005696343.2 | 2 - 3 | 649 | 105 | |
GenBank mRNA | JO409977.2 | - | 2 - 3 | 550 | 105 | |
JO427121.1 | - | 2 - 3 | 712 | 105 | ||
KA308846.1 | - | 3 - 4 | 2163 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |