Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039565.1 | XP_017895054.1 | 5 - 6 | 956 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |