Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041773.1 | XP_017897262.1 | 5 - 6 | 1123 | 94 | |
XM_018041783.1 | XP_017897272.1 | 5 - 6 | 1287 | 94 | ||
XM_018041789.1 | XP_017897278.1 | 5 - 6 | 1078 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |