Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964097.2 | XP_013819551.1 | 3 - 4 | 466 | 77 | |
GenBank mRNA | KA335133.1 | - | 6 - 7 | 1032 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |