Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056686.1 | XP_017912175.1 | 2 - 3 | 2469 | 90 | |
GenBank mRNA | JT741388.1 | - | 4 - 5 | 1606 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |