Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058699.1 | XP_017914188.1 | 4 - 5 | 1503 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |