Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060976.1 | XP_017916465.1 | 1 - 2 | 1252 | 58 | |
GenBank mRNA | JO639923.2 | - | 1 - 2 | 397 | 58 | |
KA334192.1 | - | 1 - 2 | 513 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |