Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974466.2 | XP_013829920.2 | 4 - 5 | 462 | 80 | |
XM_018039347.1 | XP_017894836.1 | 4 - 5 | 462 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |