Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO433313.2 | - | 2 - 3 | 933 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |