Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043052.1 | XP_017898541.1 | 6 - 7 | 842 | 70 | |
XM_018043053.1 | XP_017898542.1 | 6 - 7 | 940 | 70 | ||
XM_018043054.1 | XP_017898543.1 | 6 - 7 | 849 | 70 | ||
XM_018043055.1 | XP_017898544.1 | 6 - 7 | 722 | 70 | ||
GenBank mRNA | JO623382.2 | - | 6 - 7 | 961 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |