Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683885.3 | XP_005683942.3 | 2 - 3 | 927 | 70 | |
GenBank mRNA | JT649356.1 | - | 5 - 6 | 910 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |