Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692159.3 | XP_005692216.1 | 3 - 4 | 630 | 82 | |
XM_005692160.3 | XP_005692217.1 | 3 - 4 | 317 | 82 | ||
GenBank mRNA | JT678432.1 | - | 3 - 4 | 371 | 82 | |
JT702452.1 | - | 3 - 4 | 386 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |