Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062710.1 | XP_017918199.1 | 6 - 7 | 1177 | 89 | |
XM_018062711.1 | XP_017918200.1 | 6 - 7 | 1199 | 89 | ||
XM_018062712.1 | XP_017918201.1 | 6 - 7 | 1187 | 89 | ||
XM_018062713.1 | XP_017918202.1 | 6 - 7 | 1220 | 89 | ||
XM_018062714.1 | XP_017918203.1 | 6 - 7 | 1312 | 89 | ||
XM_018062717.1 | XP_017918206.1 | 6 - 7 | 1165 | 89 | ||
XM_018062718.1 | XP_017918207.1 | 6 - 7 | 1290 | 89 | ||
XM_018062719.1 | XP_017918208.1 | 5 - 6 | 1215 | 89 | ||
XM_018062720.1 | XP_017918209.1 | 5 - 6 | 1406 | 89 | ||
XM_018062721.1 | XP_017918210.1 | 5 - 6 | 908 | 89 | ||
XM_018062722.1 | XP_017918211.1 | 5 - 6 | 708 | 89 | ||
XM_018062723.1 | XP_017918212.1 | 4 - 5 | 906 | 89 | ||
XM_018062724.1 | XP_017918213.1 | 5 - 6 | 843 | 89 | ||
XM_018062725.1 | XP_017918214.1 | 5 - 6 | 618 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |