Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697562.3 | XP_005697619.2 | 3 - 4 | 720 | 150 | |
XM_005697563.3 | XP_005697620.2 | 3 - 4 | 717 | 150 | ||
GenBank mRNA | JO396540.2 | - | 1 - 2 | 832 | 150 | |
JO396541.2 | - | 1 - 2 | 832 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |