Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043532.1 | XP_017899021.1 | 15 - 16 | 1749 | 133 | |
GenBank mRNA | JT730147.1 | - | 7 - 8 | 964 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |