Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064404.1 | XP_017919893.1 | 12 - 13 | 1371 | 88 |
Gene Symbol: OVCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KA343746.1 | - | 12 - 13 | 1295 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |