Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064904.1 | XP_017920393.1 | 2 - 3 | 1325 | 118 | |
GenBank mRNA | KA315191.1 | - | 1 - 2 | 1268 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |