Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102182625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693395.2 | XP_005693452.2 | 1 | 619 | 92 |
Gene Symbol: LOC102182904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971959.2 | XP_013827413.2 | 1 | 619 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |