Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700707.3 | XP_005700764.3 | 5 - 6 | 1391 | 108 | |
GenBank mRNA | JO451805.2 | - | 1 - 2 | 492 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |