Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP36L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967250.2 | XP_013822704.2 | 2 | 718 | 106 | |
GenBank mRNA | JT710421.1 | - | 1 | 1549 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |