Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059490.1 | XP_017914979.1 | 8 | 1301 | 69 | |
GenBank mRNA | JO582076.1 | - | 1 | 2016 | 69 |
Gene Symbol: LOC102172503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696838.3 | XP_005696895.2 | 1 | 1201 | 69 | |
GenBank mRNA | JR137423.1 | - | 155 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |