Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041080.1 | XP_017896569.1 | 2 | 800 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |