Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044901.1 | XP_017900390.1 | 9 | 1234 | 58 | |
XM_018044902.1 | XP_017900391.1 | 9 | 729 | 58 | ||
XM_018044903.1 | XP_017900392.1 | 9 | 799 | 58 | ||
XM_018044904.1 | XP_017900393.1 | 9 | 726 | 58 | ||
XM_018044905.1 | XP_017900394.1 | 9 | 1189 | 58 | ||
GenBank mRNA | KA345156.1 | - | 9 | 819 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |