Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055202.1 | XP_017910691.1 | 2 | 523 | 67 | |
GenBank mRNA | JT740274.1 | - | 1 | 881 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |