Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684877.3 | XP_005684934.1 | 1 | 316 | 94 | |
XM_005684878.3 | XP_005684935.1 | 1 | 319 | 94 | ||
GenBank mRNA | JT736792.1 | - | 3 | 298 | 94 | |
KA332756.1 | - | 2 | 440 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |