Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |