Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172995.1 | NP_001166466.1 | 10 - 11 | 1759 | 84 | |
GenBank mRNA | AF004875.1 | - | 10 - 11 | 1759 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Cavia porcellus Probe spans exons |