Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173010.1 | NP_001166481.1 | 3 - 4 | 229 | 83 | |
XM_013149406.1 | XP_013004860.1 | 3 - 4 | 794 | 83 | ||
GenBank mRNA | AF484220.1 | - | 3 - 4 | 229 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |