Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003464467.4 | XP_003464515.2 | 1 - 2 | 147 | 87 | |
GenBank mRNA | AJ409054.1 | - | 1 - 2 | 36 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |