Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172940.1 | NP_001166411.1 | 3 - 4 | 1065 | 101 | |
XM_013145545.1 | XP_013000999.1 | 3 - 4 | 1098 | 101 | ||
GenBank mRNA | D11043.1 | - | 3 - 4 | 1065 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Cavia porcellus Probe spans exons |