Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173061.1 | NP_001166532.1 | 24 - 25 | 3377 | 104 | |
GenBank mRNA | D84339.1 | - | 24 - 25 | 3377 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Cavia porcellus Probe spans exons |