Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172937.2 | NP_001166408.1 | 3 - 4 | 375 | 70 | |
GenBank mRNA | M88653.1 | - | 3 - 4 | 375 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Cavia porcellus Probe spans exons |