Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173021.2 | NP_001166492.1 | 3 - 4 | 1396 | 108 | |
GenBank mRNA | X56332.1 | - | 3 - 4 | 1397 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Cavia porcellus Probe spans exons |