Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172878.2 | NP_001166349.1 | 3 - 4 | 417 | 61 | |
GenBank mRNA | AY914174.1 | - | 3 - 4 | 420 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Cavia porcellus Probe spans exons |