Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173045.1 | NP_001166516.1 | 3 - 4 | 491 | 81 | |
GenBank mRNA | FJ234418.1 | - | 3 - 4 | 491 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |