Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173073.1 | NP_001166544.1 | 2 - 3 | 191 | 103 | |
XM_005004205.3 | XP_005004262.1 | 3 - 4 | 320 | 103 | ||
XM_005004206.3 | XP_005004263.1 | 3 - 4 | 280 | 103 | ||
GenBank mRNA | EU306867.1 | - | 2 - 3 | 191 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Cavia porcellus Probe spans exons |