Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173082.1 | NP_001166553.1 | 3 - 4 | 397 | 99 | |
GenBank mRNA | EU330893.1 | - | 3 - 4 | 397 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Cavia porcellus Probe spans exons |