Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173122.1 | NP_001166593.1 | 7 - 8 | 1141 | 70 | |
XM_013146782.1 | XP_013002236.1 | 7 - 8 | 1189 | 70 | ||
GenBank mRNA | DQ911473.1 | - | 7 - 8 | 1141 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Cavia porcellus Probe spans exons |