Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Std1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172893.1 | NP_001166364.1 | 5 - 6 | 786 | 92 | |
GenBank mRNA | U06871.1 | - | 5 - 6 | 786 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Cavia porcellus Probe spans exons |