Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173401.1 | NP_001166872.1 | 3 - 4 | 611 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |