Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172740.1 | NP_001166211.1 | 7 - 8 | 1007 | 77 | |
GenBank mRNA | AJ009673.1 | - | 7 - 8 | 1007 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Cavia porcellus Probe spans exons |