Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172755.1 | NP_001166226.1 | 1 - 2 | 1328 | 61 | |
GenBank mRNA | AJ534845.1 | - | 1 - 2 | 1328 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Cavia porcellus Probe spans exons |