Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172762.1 | NP_001166233.1 | 2 - 3 | 714 | 82 | |
GenBank mRNA | AY075099.1 | - | 2 - 3 | 714 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |