Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172836.1 | NP_001166307.1 | 7 - 8 | 696 | 58 | |
XM_013142665.2 | XP_012998119.1 | 7 - 8 | 978 | 58 | ||
GenBank mRNA | AB006444.1 | - | 7 - 8 | 696 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Cavia porcellus Probe spans exons |