Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173087.1 | NP_001166558.1 | 2 - 3 | 192 | 81 | |
GenBank mRNA | DQ324462.1 | - | 2 - 3 | 192 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |