Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273505.1 | NP_001260434.1 | 4 - 5 | 1793 | 69 | |
NM_135767.2 | NP_609611.1 | 4 - 5 | 1793 | 69 | ||
GenBank mRNA | BT056246.1 | - | 4 - 5 | 1790 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |