Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loqs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038814.3 | NP_001033903.1 | 2 - 3 | 909 | 65 | |
NM_001201867.2 | NP_001188796.1 | 2 - 3 | 909 | 65 | ||
NM_001298964.1 | NP_001285893.1 | 2 - 3 | 909 | 65 | ||
NM_001298965.1 | NP_001285894.1 | 2 - 3 | 909 | 65 | ||
NM_135802.4 | NP_609646.1 | 2 - 3 | 909 | 65 | ||
NM_165041.3 | NP_723813.1 | 2 - 3 | 909 | 65 | ||
GenBank mRNA | AY071070.1 | - | 2 - 3 | 906 | 65 | |
DQ026059.1 | - | 1 - 2 | 729 | 65 | ||
DQ118034.1 | - | 2 - 3 | 913 | 65 | ||
DQ118035.1 | - | 2 - 3 | 913 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |